Learn More
Bio Rad Laboratories LUER TO 10-32 ADAPTOR FITTINGS
Catalog No. 509011074
Encompass
Kit, Adaptor; Bio-Rad; Accessories for use w/ Bio-Scale Mini cartridges; Includes: Luer to 10-32 fittings to connect 1 cartridge to HPLC system
Catalog No. 50-901-1074 Supplier Bio Rad Laboratories Supplier No. 7320112
Kit, Adaptor; Bio-Rad; Accessories for use w/ Bio-Scale Mini cartridges; Includes: Luer to 10-32 fittings to connect 1 cartridge to HPLC system