Learn More
Bio Rad Laboratories LUER TO BIOLOGIC SYSTEM FITTIN
Catalog No. 509011075
Encompass
Kit, Fittings; Bio-Rad; Accessories for use w/ Bio-Scale Mini Cartridges; Includes: 0.25 to 28 female to male luer, 0.25 to 28 female to female luer, to connect 1 Bio-Scale Mini cartridge to a BioLogic DuoFlow system
Catalog No. 50-901-1075 Supplier Bio Rad Laboratories Supplier No. 7320113
Kit, Fittings; Bio-Rad; Accessories for use w/ Bio-Scale Mini Cartridges; Includes: 0.25 to 28 female to male luer, 0.25 to 28 female to female luer, to connect 1 Bio-Scale Mini cartridge to a BioLogic DuoFlow system