Learn More
Sigma Aldrich Fine Chemicals Biosciences Luer-to-Tubing Connector b1EA
Catalog No. 502900119
Encompass
Hardware specifically designed for secure leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. 316 SS fittings are noncontaminating easy to clean and resistant to corrosion.Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Catalog No. 50-290-0119 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2614911EA
Hardware specifically designed for secure leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. 316 SS fittings are noncontaminating easy to clean and resistant to corrosion.Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.