Learn More
Sigma Aldrich Fine Chemicals Biosciences Luer-to-Tubing Connector m
Catalog No. 502900647
Encompass
Hardware specifically designed for secure leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. 316 SS fittings are noncontaminating easy to clean and resistant to corrosion.Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Catalog No. 50-290-0647 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1180284EA
Hardware specifically designed for secure leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. 316 SS fittings are noncontaminating easy to clean and resistant to corrosion.Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.