Learn More
World Precision Instrument Luer Valve Assortment Kit
SDP
Catalog No. 50822019
Encompass
Build your own liquid flow experiment with this Luer kit
This Luer fittings kit is useful for building your own liquid flow experiment. It provides the means to start, stop, add, divide and control a flow of liquid or gas. Included in the kit are over 300 as­sort­ed parts such as one-way and 3-way stopcocks, manifolds, Y-connectors, in­jec­tion sites, male and female luer caps, check valves, syringe-activated check valves, slide clamps, roller clamps, and pinch clamps. All (except clamps) have a luer fitting for quick and easy con­nect­ing and disconnecting. This Luer fittings kit ncludes assorted luer fittings for use with flex­i­ble tubing.
Catalog No. 50-822-019 Supplier World Precision Instrument Supplier No. 14011
 Restricted Item
Add to Cart
Edge
Add to Cart
Build your own liquid flow experiment with this Luer kit
This Luer fittings kit is useful for building your own liquid flow experiment. It provides the means to start, stop, add, divide and control a flow of liquid or gas. Included in the kit are over 300 as­sort­ed parts such as one-way and 3-way stopcocks, manifolds, Y-connectors, in­jec­tion sites, male and female luer caps, check valves, syringe-activated check valves, slide clamps, roller clamps, and pinch clamps. All (except clamps) have a luer fitting for quick and easy con­nect­ing and disconnecting. This Luer fittings kit ncludes assorted luer fittings for use with flex­i­ble tubing.