Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polycarbonate, Straight, Male Luer Lock to Low-Profile Hose Barb Adapter, 1/16"" ID; 25/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4550407
Encompass
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring. Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
Catalog No. NC1108506
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.