Learn More
AVANTOR MASTERFLEX (VWR LLC) Male luer with lock ring x 3/32"" hose barb, PP, 25/pk
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4551802
Encompass
Male Luer with lock ring x 3/32" hose barb, PP, 25/pkFull material traceability to ensure process integrityManufactured from 100% virgin raw materials in a Class 100,000 environmentThese luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of options including luer, thread, and hose barb.
Catalog No. NC0445352