Learn More
Ibidi USA Luer Lock Connector Male
SDP
Supplier:  Ibidi USA 10826
Encompass
Luer Lock Connector Male connects female Luers with 0.5, 0.8, or 1.6mm ID tubing. polypropylene, sterile, 25 pcs./box
Catalog No. NC0700581