Learn More
Andwin Scientific LUERLOCK SYRINGEW/O NEED/PK100
SDP
Catalog No. NC0769627
Encompass_Preferred
The Norm-Ject Syringe is made with all plastic, polypropylene. This product is suitable to use with sharp, tri-bevel needles. The syringes are certified for use with most common organic andinorganic chromatography solvents. This does not have any latex, BPA, styrene, DEHP, rubber or silicone that could pollute the chromatography or sensitive laboratory samples. It is sold in a pack of 100.
Catalog No. NC0769627 Supplier Andwin Scientific Supplier No. 4020.X00V0
The Norm-Ject Syringe is made with all plastic, polypropylene. This product is suitable to use with sharp, tri-bevel needles. The syringes are certified for use with most common organic andinorganic chromatography solvents. This does not have any latex, BPA, styrene, DEHP, rubber or silicone that could pollute the chromatography or sensitive laboratory samples. It is sold in a pack of 100.