Learn More
Organomation LUERS/WASHERS F/N-EVAP 12/PK
SDP
Catalog No. NC2599928
Encompass_Preferred
The luer and washer attaches to the valve tube of all N-EVAPs to allow for the connection of needles. The luer and washer is also used on the gas manifolds of the 15 position MICROVAP and the 24 position MICROVAP. Each unit contains 12 luer and washers.Specifications:#10-32 threads to attach luer to valve tubeFemale luer with plastic washerChrome plated brass
Catalog No. NC2599928 Supplier Organomation Supplier No. P060712
The luer and washer attaches to the valve tube of all N-EVAPs to allow for the connection of needles. The luer and washer is also used on the gas manifolds of the 15 position MICROVAP and the 24 position MICROVAP. Each unit contains 12 luer and washers.Specifications:#10-32 threads to attach luer to valve tubeFemale luer with plastic washerChrome plated brass