Learn More
Medchemexpress LLC Luf5771 | 1141802-49-4 | MFCD34567719 | 99.5% | 357.44 g/mol | C24H23NO2 | 5 MG
SDP
Catalog No. 5000221006
Encompass_Preferred
LUF5771 is a small-molecule allosteric modulator that inhibits recombinant luteinizing hormone (recLH) and Org 43553 activity while partially activating the LH receptor with low efficacy. Supplied for research use, it is offered as a high-purity solid and as a DMSO solution for in vitro receptor pharmacology studies.
  • Potent allosteric inhibitor of recLH and Org 43553.
  • Partial agonist activity at the LH receptor with low efficacy.
  • High purity (about 99.5%).
  • Available as solid powder and 10 mM solution in DMSO.
  • Suitable for in vitro receptor pharmacology and assay development.
  • Molecular formula C24H23NO2; molecular weight 357.44 g/mol.
Catalog No. 50-002-21006 Supplier Medchemexpress LLC Supplier No. HY1393035MG
LUF5771 is a small-molecule allosteric modulator that inhibits recombinant luteinizing hormone (recLH) and Org 43553 activity while partially activating the LH receptor with low efficacy. Supplied for research use, it is offered as a high-purity solid and as a DMSO solution for in vitro receptor pharmacology studies.
  • Potent allosteric inhibitor of recLH and Org 43553.
  • Partial agonist activity at the LH receptor with low efficacy.
  • High purity (about 99.5%).
  • Available as solid powder and 10 mM solution in DMSO.
  • Suitable for in vitro receptor pharmacology and assay development.
  • Molecular formula C24H23NO2; molecular weight 357.44 g/mol.