Learn More
Medchemexpress LLC HY-13236 5mg Medchemexpress, LUF6000 CAS:890087-21-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY132365MG
Encompass_Preferred
Medchemexpress, HY-13236 5mg LUF6000 CAS:890087-21-5 LUF6000 is an orally active allosteric modulator of the A3 adenosine receptor. LUF6000 has potent anti-inflammatory effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1310