Learn More
Medchemexpress LLC LUF6096 | 1116652-18-6 | 99.16% | 414.33 | 100 MG
SDP
Catalog No. 5000344283
Encompass_Preferred
LUF6096 is a potent allosteric enhancer of the adenosine A3 receptor, capable of enhancing agonist binding. It exhibits low orthosteric affinity for any of the adenosine receptors and demonstrates protective effects in myocardial ischemia/reperfusion injury.
  • Enhances agonist binding of the adenosine A3 receptor.
  • Exhibits low orthosteric affinity for adenosine receptors.
  • Shows protective effects in myocardial ischemia/reperfusion injury.
  • Available in various sizes for research needs.
  • High purity for reliable experimental results.
Catalog No. 50-003-44283 Supplier Medchemexpress LLC Supplier No. HY10915100MG
LUF6096 is a potent allosteric enhancer of the adenosine A3 receptor, capable of enhancing agonist binding. It exhibits low orthosteric affinity for any of the adenosine receptors and demonstrates protective effects in myocardial ischemia/reperfusion injury.
  • Enhances agonist binding of the adenosine A3 receptor.
  • Exhibits low orthosteric affinity for adenosine receptors.
  • Shows protective effects in myocardial ischemia/reperfusion injury.
  • Available in various sizes for research needs.
  • High purity for reliable experimental results.