Learn More
Medchemexpress LLC Luf6096 | 1116652-18-6 | 99.2% | 414.33 | 10 MG
SDP
Catalog No. 5000344782
Encompass_Preferred
LUF6096 is a potent allosteric enhancer of the adenosine A3 receptor, which can allosterically enhance agonist binding. It exhibits low orthosteric affinity for any of the adenosine receptors. LUF6096 has shown protective effects in myocardial ischemia/reperfusion injury.
  • Potent allosteric enhancer of adenosine A3 receptor
  • Exhibits low orthosteric affinity for adenosine receptors
  • Decreases dissociation rate of 125I-AB-MECA from A3 receptor
  • Enhances intrinsic activity of Cl-IB-MECA for inhibition of forskolin-stimulated cAMP production
  • Protects against myocardial ischemia/reperfusion injury in dogs
  • Reduces infarct size in animal models
Catalog No. 50-003-44782 Supplier Medchemexpress LLC Supplier No. HY1091510MG
LUF6096 is a potent allosteric enhancer of the adenosine A3 receptor, which can allosterically enhance agonist binding. It exhibits low orthosteric affinity for any of the adenosine receptors. LUF6096 has shown protective effects in myocardial ischemia/reperfusion injury.
  • Potent allosteric enhancer of adenosine A3 receptor
  • Exhibits low orthosteric affinity for adenosine receptors
  • Decreases dissociation rate of 125I-AB-MECA from A3 receptor
  • Enhances intrinsic activity of Cl-IB-MECA for inhibition of forskolin-stimulated cAMP production
  • Protects against myocardial ischemia/reperfusion injury in dogs
  • Reduces infarct size in animal models