Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LUF6096 | 1116652-18-6 | 99.2% | 414.33 g/mol | C22H21Cl2N3O | 5 MG
SDP

Catalog No. 5000264726
Encompass_Preferred

LUF6096 is a small-molecule allosteric enhancer of the adenosine A3 receptor supplied as a purified solid for research use. It enhances agonist binding to A3AR and includes recommended storage and formulation guidance for in vitro and in vivo studies.

  • Allosteric enhancer of adenosine A3 receptor
  • High purity: 99.16%
  • Available in small research packs, including 5 mg
  • Molecular weight 414.33 g/mol; formula C22H21Cl2N3O
  • Recommended in vitro solvent: DMSO (20 mg/mL); in vivo formulation guidance provided
  • Storage: powder at -20°C; in solvent store at -80°C for extended periods

Catalog No. 50-002-64726 Supplier Medchemexpress LLC Supplier No. HY109155MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LUF6096 is a small-molecule allosteric enhancer of the adenosine A3 receptor supplied as a purified solid for research use. It enhances agonist binding to A3AR and includes recommended storage and formulation guidance for in vitro and in vivo studies.

  • Allosteric enhancer of adenosine A3 receptor
  • High purity: 99.16%
  • Available in small research packs, including 5 mg
  • Molecular weight 414.33 g/mol; formula C22H21Cl2N3O
  • Recommended in vitro solvent: DMSO (20 mg/mL); in vivo formulation guidance provided
  • Storage: powder at -20°C; in solvent store at -80°C for extended periods

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.