Learn More
Medchemexpress LLC Lufenuron 100mg | 103055-07-8 | 511.1 g/mol | C17H8Cl2F8N2O3 | 100 MG
SDP
Catalog No. 5000229412
Encompass_Preferred
Lufenuron is a benzoylurea insecticide and chitin synthesis inhibitor supplied for laboratory research. This research-grade material is provided as a small-scale, high-purity solid intended for experimental and analytical use; it is not for human or veterinary applications.
  • High purity suitable for analytical and experimental work.
  • Packaged as a 100 mg solid for small-scale studies.
  • CAS number 103055-07-8 for unambiguous identification.
  • Available COA, SDS, and data sheet for quality and safety documentation.
  • Lipophilic benzoylurea class compound acting as a chitin synthesis inhibitor.
Catalog No. 50-002-29412 Supplier Medchemexpress LLC Supplier No. HY115584100MG
Lufenuron is a benzoylurea insecticide and chitin synthesis inhibitor supplied for laboratory research. This research-grade material is provided as a small-scale, high-purity solid intended for experimental and analytical use; it is not for human or veterinary applications.
  • High purity suitable for analytical and experimental work.
  • Packaged as a 100 mg solid for small-scale studies.
  • CAS number 103055-07-8 for unambiguous identification.
  • Available COA, SDS, and data sheet for quality and safety documentation.
  • Lipophilic benzoylurea class compound acting as a chitin synthesis inhibitor.