Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luff-Schoorl Reagent, Solstice

Catalog No. 6000912 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-009-12 1 L
60-009-13 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-009-12 Supplier Solstice Supplier No. 344001L
Add to Cart
Add to Cart

Volumetric, 25 g/L

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Luff-Schoorl Reagent
CAS 497-19-8, 7732-18-5, 7758-98-7
UN Number UN3082
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.