Learn More
Grainger Eaton FAZ Feed Extension Terminal Lug, Single-Pole, 80 A, 1 ea
Catalog No. 502795255
Encompass_Preferred
The Eaton FAZ feed extension terminal lug is a single-pole accessory designed for use with miniature circuit breakers. This feed-in terminal is engineered to facilitate power distribution to compatible busbars in industrial and commercial electrical systems.
  • Amperage rating of 80 A
  • Voltage rating of 480 VAC
  • Compatible with conductor sizes ranging from #14 to #2 AWG
  • Box-type lug configuration
  • UL 489 certified
  • IP20 protection rating
  • Copper construction
Catalog No. 50-279-5255 Supplier Grainger Supplier No. 19YH01
The Eaton FAZ feed extension terminal lug is a single-pole accessory designed for use with miniature circuit breakers. This feed-in terminal is engineered to facilitate power distribution to compatible busbars in industrial and commercial electrical systems.
  • Amperage rating of 80 A
  • Voltage rating of 480 VAC
  • Compatible with conductor sizes ranging from #14 to #2 AWG
  • Box-type lug configuration
  • UL 489 certified
  • IP20 protection rating
  • Copper construction