Learn More
Volu Sol Lugol's Iodine (Diluent Included), 8 mL & 16 oz / 500 mL, Volu-Sol™
SDP
Catalog No. 50420801 Shop All Volu Sol Products
Encompass_Preferred
Two-part solution that includes a Lugol's solution and a dilutent.  Can be used in Gram staining methods.  Also used to test for the presence of starch in organic compounds
This item is not returnable. View return policy
Catalog No. 50-420-801 Supplier Volu Sol Supplier No. VIG016
Two-part solution that includes a Lugol's solution and a dilutent.  Can be used in Gram staining methods.  Also used to test for the presence of starch in organic compounds