Learn More
Volu Sol Lugol's Iodine (Diluent Included), 4 mL & 8 oz / 250 mL, Volu-Sol™
SDP
Catalog No. 50420800 Shop All Volu Sol Products
Encompass_Preferred
Two-part solution that includes a Lugol's solution and a dilutent.  Can be used in Gram staining methods.  Also used to test for the presence of starch in organic compounds
Catalog No. 50-420-800 Supplier Volu Sol Supplier No. VIG008
Two-part solution that includes a Lugol's solution and a dilutent.  Can be used in Gram staining methods.  Also used to test for the presence of starch in organic compounds