Learn More
ENG Scientific Lugol's Iodine Solution
Catalog No. ES6860
Change view
Click to view available options
Quantity:
2 x 950 mL
Catalog No. Quantity
ES6860 2 x 950 mL
1 options
Catalog No. ES6860 Supplier ENG Scientific Supplier No. 6860
Add to Cart
Add to Cart
For the observation of protozoan cysts in wet mount preparations of fecal specimens. For Lugol's Iodine Solution. Includes Iodine, potassium iodide.
  • Used primarily for the observation of protozoan cysts in wet mount preparations of fecal specimens.
  • Results: Iodine enhances the nuclear structure and stains glycogen masses dark brown.
  • Lugol's Iodine Solution Kit
  • Includes Iodine, potassium iodide
  • Bottles
Specifications
Chemical Name or Material Lugol's Iodine Solution
Packaging Bottles
Quantity 2 x 950 mL
For in vitro diagnostic use only.