Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lugol's solution

Catalog No. p-7078296 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1L
2.5L
Packaging Type:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
M1005672500 2.5L Plastic Bottle
M1005671000 1L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1005672500 Supplier MilliporeSigma™ Supplier No. 1.00567.2500
Only null left
Add to Cart
Add to Cart

Used in bacteriological staining Gram bacteria.

  • Contains complex of Iodine-Polyvinylpyrrolidon (PVP)
  • No free iodine, which stabilizes the solution and reduces the evaporation of iodine
  • Lugol’s solution and crystal violet form stable complex that will not be released by decolorisation
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2.5L
Storage Requirements 15° to 25°C
Packaging Type Plastic Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.