Learn More
Medchemexpress LLC Luliconazole | 187164-19-8 | 99.95% | C14H9Cl2N3S2 | 250 MG
SDP
Catalog No. 5000352520
Encompass_Preferred
Luliconazole (NND 502) is a topical antifungal imidazole antibiotic with broad-spectrum and potent antifungal activity. It is a solid, white to yellow in color.
  • Can be used for the research of skin infections, including dermatophytosis, tinea corporis, and tinea pedis.
  • Shows great potency against Trichophyton spp. (MIC range: 0.00012-0.002 μg/ml).
  • Highly active against Candida albicans (MIC range: 0.031-0.13 μg/ml).
  • Very low MIC against Malassezia restricta (MIC range: 0.004-0.016 μg/ml).
  • Administered topically, it shows dose-dependent therapeutic efficacy on skin for dermatophytosis.
Catalog No. 50-003-52520 Supplier Medchemexpress LLC Supplier No. 250MGHY14283
Luliconazole (NND 502) is a topical antifungal imidazole antibiotic with broad-spectrum and potent antifungal activity. It is a solid, white to yellow in color.
  • Can be used for the research of skin infections, including dermatophytosis, tinea corporis, and tinea pedis.
  • Shows great potency against Trichophyton spp. (MIC range: 0.00012-0.002 μg/ml).
  • Highly active against Candida albicans (MIC range: 0.031-0.13 μg/ml).
  • Very low MIC against Malassezia restricta (MIC range: 0.004-0.016 μg/ml).
  • Administered topically, it shows dose-dependent therapeutic efficacy on skin for dermatophytosis.