Learn More
Boca Scientific Inc Lullaby siRNA Transfection Reagent, 500µl (1000 assays)
Supplier:  Boca Scientific Inc LL70500
Encompass
Lullaby is the ideal siRNA transfection reagent for gene silencing. This reagent uses TEE Technology (Triggered Endosomal Escape), a mechanism that combines and exploits the properties of cationic lipids and polymers to achieve extremely efficient DNA delivery into cells. Lullaby has been successfully tested on numerous cell lines, reaching up to 90% gene silencing with high reproducibility and very low toxicity. RNA interference is a powerful technique to shut down gene expression in cells and organisms. Short RNA duplexes are extremely selective by interacting and inducing the degradation of their specific mRNA targets, and thereby inhibit the resulting protein production. Lullaby siRNA Transfection Reagent introduces the siRNA duplexes in a variety of cells with a very high efficiency, leading to exceptional knockdown effects with low doses of siRNA.
Catalog No. NC1113822