Learn More
Medchemexpress LLC HY-13262 10mg Medchemexpress, Lumacaftor CAS:936727-05-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1326210MG
Encompass_Preferred
Medchemexpress, HY-13262 10mg Lumacaftor CAS:936727-05-8 Lumacaftor (VX-809; VRT 826809) is a CFTR modulator that corrects the folding and trafficking of CFTR protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8974