Learn More
Medchemexpress LLC Lumacaftor-d4 | 2733561-44-7 | MFCD00143661 | 99.8% | 456.43 | C24H14D4F2N2O5 | 5 MG
SDP
Catalog No. 5000262736
Encompass_Preferred
Lumacaftor-d4 is the deuterium-labeled analogue of lumacaftor supplied for research applications. It is intended as a tracer and internal standard to support accurate, quantitative analyses by NMR, GC-MS, and LC-MS in laboratory settings.
  • Deuterium-labeled internal standard for quantitative analysis by MS and NMR.
  • High purity suitable for analytical and tracer applications.
  • Available in small-mass packages for trace-level work.
  • Stable isotope labeling improves accuracy and precision in quantitation.
  • Intended for research use only; not for clinical or therapeutic use.
Catalog No. 50-002-62736 Supplier Medchemexpress LLC Supplier No. HY13262S5MG
Lumacaftor-d4 is the deuterium-labeled analogue of lumacaftor supplied for research applications. It is intended as a tracer and internal standard to support accurate, quantitative analyses by NMR, GC-MS, and LC-MS in laboratory settings.
  • Deuterium-labeled internal standard for quantitative analysis by MS and NMR.
  • High purity suitable for analytical and tracer applications.
  • Available in small-mass packages for trace-level work.
  • Stable isotope labeling improves accuracy and precision in quantitation.
  • Intended for research use only; not for clinical or therapeutic use.