Learn More
Revvity Health Sciences Inc LumaPlate-96 DeepWell, White Opaque 96-well Microplate with Scintillant Coated on the Bottom, hold up to 300 µl
Catalog No. 502810302
Encompass
LumaPlate-96 DeepWell, White Opaque 96-well Microplate with Scintillant Coated on the Bottom, hold up to 300 µl
Catalog No. 50-281-0302 Supplier Revvity Health Sciences Inc Supplier No. 6065630
LumaPlate-96 DeepWell, White Opaque 96-well Microplate with Scintillant Coated on the Bottom, hold up to 300 µl