Learn More
Selleck Chemical LLC Lumateperone Tosylate E4874-100MG
SDP
Catalog No. 5000041770
Encompass_Preferred
Lumateperone tosylate(ITI-007) is an antagonist of 5-HT2A receptor It also acts as a partial agonist of presynaptic D2 receptors as an antagonist of postsynaptic D2 receptors and as a modulator of dopamine D1 receptor Lumateperone tosylate demonstrates anticancer activity
Catalog No. 50-000-41770 Supplier Selleck Chemical LLC Supplier No. E4874100MG
Lumateperone tosylate(ITI-007) is an antagonist of 5-HT2A receptor It also acts as a partial agonist of presynaptic D2 receptors as an antagonist of postsynaptic D2 receptors and as a modulator of dopamine D1 receptor Lumateperone tosylate demonstrates anticancer activity