Learn More
Mopec Lumea Anti-Slip Platform
SDP
Catalog No. 22024813 Shop All Mopec Products
Change view
Click to view available options
Color:
Pink
Catalog No. Color
22-024-813 Pink
1 options
Catalog No. 22-024-813 Supplier Mopec Supplier No. BX111
Add to Cart
Add to Cart
Provides a steady, secure base for your BxBoard, helping it stay firmly in place while handling biopsy samples
  • Provides a stable, non-slip base for the BxBoard.
  • Helps prevent shifting while loading biopsy samples.
  • Improves precision and control during specimen handling.
  • Useful for procedures performed without an assistant.
  • The Anti-Slip Platform stabilizes the BxBoard™ during tissue loading, preventing unwanted movement or sliding.
  • This ensures safer handling for the user and helps preserve the integrity of the patient tissue.
  • Mopec is the exclusive supplier of Lumea consumable products.
  • Durable, easy-to-clean design for repeated clinical use.
Specifications
Color Pink