Learn More
Mopec Lumea BxBoard Prostate Kit
SDP
Catalog No. 22024808 Shop All Mopec Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
22-024-808 Polypropylene
1 options
Catalog No. 22-024-808 Supplier Mopec Supplier No. BX106
Add to Cart
Add to Cart
Small tissue, 6-lane Universal BxBoard™
  • 1 BxBoard for small tissue with 6-lanes
  • Hydrophilic formalin-soaked sponge technology preserves anatomic orientation of biopsy tissue
  • Designed to reduce formaldehyde exposure for medical assistants, physicians, and laboratory personnel
  • Clinics report an 83% reduction in specimen containers and an 89% reduction in formalin per prostate case
  • Provides an overall 136% reduction in biohazard waste per histology lab
  • Mopec is the exclusive supplier of Lumea™ consumable products
Specifications
Sterility Non-sterile
Color Black
Tip Material Sponge
Material Polypropylene