Learn More
Mopec Lumea BxBoard Prostate Kit, Transrectal, 6 Lane, Left/Right
SDP
Catalog No. 22024811 Shop All Mopec Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
22-024-811 Polypropylene
1 options
Catalog No. 22-024-811 Supplier Mopec Supplier No. BX109
Add to Cart
Add to Cart
Core Needle Biopsy, 6-lane prostate BxBoard™
  • Includes
    • 1 6-Site Left Green BxBoard lat base, base, lat mid, mid, lat apex, apex
    • 1 6-Site Right Blue BxBoard base, lat base, mid, lat mid, apex, lat apex
    • 1 Prostate Biopsy Corrections/Additional Samples Form
    • 1 Protective Transport Mailer Box 6.5 x 6 x 2 in.
  • Hydrophilic formalin-soaked sponge technology preserves anatomic orientation of biopsy tissue.
  • Designed to reduce formaldehyde exposure for medical assistants, physicians, and laboratory personnel.
  • Clinics report an 83% reduction in specimen containers and an 89% reduction in formalin per prostate case.
  • Provides an overall 136% reduction in biohazard waste per histology lab.
  • Mopec is the exclusive supplier of Lumea™ consumable products.
Specifications
Sterility Non-sterile
Color Blue
Tip Material Sponge
Material Polypropylene