Learn More
Mopec Lumea BxBoard Prostate Kit, Transrectal, 6 Lane, Left/Right, Blank
SDP
Catalog No. 22024810 Shop All Mopec Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
22-024-810 Polypropylene
1 options
Catalog No. 22-024-810 Supplier Mopec Supplier No. BX108
Add to Cart
Add to Cart
Designed to preserve anatomic orientation while reducing formaldehyde exposure and waste.
  • Includes:
    • 1 - 6-Site Left (Green) BxBoard (lat base, base, lat mid, mid, lat apex, apex)
    • 1 - 6-Site Right (Blue) BxBoard (base, lat base, mid, lat mid, apex, lat apex)
    • 1 - X1 Outlier (Black) BxBoard
    • 1 - Transrectal Biopsy Corrections/Additional Samples Form
    • 1 - Protective Transport Mailer Box (6.5 x 5.5 x 3.75 in.)
  • Hydrophilic formalin-soaked sponge technology preserves anatomic orientation of biopsy tissue.
  • Designed to reduce formaldehyde exposure for medical assistants, physicians, and laboratory personnel.
  • Clinics report an 83% reduction in specimen containers and an 89% reduction in formalin per prostate case.
  • Provides an overall 136% reduction in biohazard waste per histology lab.
  • Mopec is the exclusive supplier of Lumea™ consumable products.
Specifications
Sterility Non-sterile
Color Blue
Tip Material Sponge
Material Polypropylene