Learn More
Mopec Lumea BxBoard Prostate Transperineal Kit
SDP
Catalog No. 22024812 Shop All Mopec Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
22-024-812 Polypropylene
1 options
Catalog No. 22-024-812 Supplier Mopec Supplier No. BX110
Add to Cart
Add to Cart
4-board BxBoard Transperineal Prostate Biopsy Kit. Lumea™ BxLink™ compatible.
  • Includes:
    • 1 Left 1-6 Green BxBoard
    • 1 Left 7-12 Green BxBoard
    • 1 Right 1-6 Blue BxBoard
    • 1 Right 7-12 Blue BxBoard
    • 1 TPN Biopsy Corrections Additional Samples Form
    • 1 Protective Transport Mailer Box 6.5 x 5.5 x 3.75 in.
  • Hydrophilic formalin-soaked sponge technology preserves anatomic orientation of biopsy tissue
  • Designed to reduce formaldehyde exposure for medical assistants, physicians, and laboratory personnel
  • Clinics report an 83 reduction in specimen containers and an 89 reduction in formalin per prostate case
  • Provides an overall 136 reduction in biohazard waste per histology lab
  • Mopec is the exclusive supplier of Lumea™ consumable products
Specifications
Sterility Non-sterile
Color Blue
Tip Material Sponge
Material Polypropylene