Learn More
Mopec Lumea BxCamera Software Subscription
SDP
Catalog No. 21402124 Shop All Mopec Products
Change view
Click to view available options
Includes:
1 Year Service
2 Year Service
3 Year Service
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-402-124 1 Year Service
21-402-125 2 Year Service
21-402-126 3 Year Service
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-402-124 Supplier Mopec Supplier No. SW100
Only null left
Add to Cart
Add to Cart
Software guides users through setup and operation with intuitive controls for image capture and automated biopsy measurement. Supports standardized reporting, real-time feedback, and workflow integration. Requires a valid software subscription for continued use.
  • Supports automatic measurement and documentation of small biopsy tissue
  • Provides real-time measurement feedback and structured gross description outputs
  • Includes tools for connecting captured data to other lab systems
  • Reduces grossing time and improves data consistency
  • Supports quality assurance in pathology processes
Specifications
Product Type Software Subscription
Includes 1 Year Service
For Use With (Equipment) Lumea BxCamera
Control Type Digital