Description
- Supports automatic measurement and documentation of small biopsy tissue
- Provides real-time measurement feedback and structured gross description outputs
- Includes tools for connecting captured data to other lab systems
- Reduces grossing time and improves data consistency
- Supports quality assurance in pathology processes
Specifications
Specifications
| Product Type | Software Subscription |
| Includes | 3 Year Service |
| For Use With (Equipment) | Lumea BxCamera |
| Control Type | Digital |