Learn More
Mopec Lumea BxChip For Prostate
SDP
Catalog No. 22024803 Shop All Mopec Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
22-024-803 Blue
1 options
Catalog No. 22-024-803 Supplier Mopec Supplier No. BX101
Add to Cart
Add to Cart
Small-tissue 6-lane chip securely holds multiple biopsy specimens in one plane through processing for consistent histologic results.
  • Small tissue, 6-lane BxChip™ format, with 50 chips supplied per bottle.
  • Designed to securely retain multiple biopsy specimens collected directly from a specimen container or from the BxBoard™.
  • Maintains tissue orientation in a single plane throughout fixation, processing, embedding, and slide preparation to ensure consistency and specimen integrity.
  • Reduces common issues seen with traditional embedding methods, such as tissue curling, overlap, and fragmentation of biopsy cores.
  • Improved consistency has led some pathologists to report higher cancer detection rates when using the BxChip™.
  • Manufactured from a specialized material that mimics human tissue behavior during histological processing.
  • Engineered to shrink slightly during processing to firmly hold specimens in place and allow clean sectioning.
  • Chip material lightly stains on the slide, providing a background reference while preserving original tissue core positioning.
  • Includes embossed markings for accurate core tracking and orientation.
  • Directional arrows indicate proximal and distal ends of tissue samples.
  • Unique dot patterns identify each core position from left to right.
  • Supports both manual and software-based core identification, reducing errors and enhancing diagnostic clarity.
  • When used with Lumea's BxCamera™ and software integrated into the Mopec Maestro™ Encore Grossing Station, tissue placement and labeling are automatically tracked.
  • Eliminates the need for manual documentation through automated tracking.
  • Software enables pathologists to designate areas of interest on digital slides, automatically calculate percentage involvement, and generate a draft diagnostic report.
  • Mopec is the exclusive supplier of Lumea consumable products.
Specifications
Color Blue
Disposable Single-use
Size Small