Learn More
Mopec Lumea BxTamper
SDP
Catalog No. 22024806 Shop All Mopec Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
22-024-806 Black
1 options
Catalog No. 22-024-806 Supplier Mopec Supplier No. BX104
Add to Cart
Add to Cart
Lumea embedding tampers are precisely engineered to apply uniform pressure across the entire surface of the BxChip™.
  • Embedding tampers used to flatten tissue sections within embedding molds, ensuring proper orientation and consistency. Includes an aluminum base.
  • Lumea's embedding tampers are precisely engineered to apply uniform pressure across the entire surface of the BxChip™.
  • Featuring an aluminum base, the tamper heats quickly and resists sticking to the back of the chip.
  • Its ergonomic handle is designed to fit comfortably between the thumb and index finger, minimizing hand fatigue during repeated use.
  • Mopec is the exclusive supplier of Lumea consumable products.
Specifications
Color Black