Learn More
Mopec Lumea Lab Demo Kit
SDP
Catalog No. 22024804 Shop All Mopec Products
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
22-024-804 Non-sterile
1 options
Catalog No. 22-024-804 Supplier Mopec Supplier No. BX102
Add to Cart
Add to Cart
Lumea, BxChip For Prostate, 6 Lane, Lab Demo Kit
  • 1 BxBoard™ (6 lane) loaded with porcine needle cores.
  • 1 bottle of 10 unprocessed BxChips.
  • 1 bag of 10 white and 10 blue sponges (used for processing 10 unprocessed BxChips).
  • 1 cassette that shows where to place the sponges (white on bottom, blue on top).
  • 2 FFPE blocks with BxChips loaded with porcine needle cores (to practice microtomy).
  • 2 processed cassettes with tamper tool (to practice embedding).
  • A small card with a link to our website for instructions.
  • The BxChip™ is perfect for medical laboratories.
  • Patented Sectionable Matrix technology allows increased tissue surface area and preserves the integrity of the tissue.
  • Aids pathologists in cancer detection rate.
  • Mopec is the exclusive supplier of Lumea consumable products.
Specifications
Product Type Specimen Collection Kit
Sterility Non-sterile