Learn More
Medchemexpress LLC Lumefantrine-d9 | 2477594-24-2 | 97.1% | 538.00 | C30H23D9Cl3NO | 10 MG
SDP
Catalog No. 5000296698
Encompass_Preferred
Lumefantrine-d9 is a deuterium-labeled analog of lumefantrine used as an isotopically labeled internal standard and reference for analytical, pharmacokinetic, and metabolic studies. It enables accurate quantitation and method validation in mass spectrometry workflows.
  • Deuterium-labeled internal standard for lumefantrine assays.
  • Suitable for mass spectrometry based quantitation and tracer studies.
  • High purity and characterized molecular weight for reliable analytical results.
  • Stable as a powder; recommended storage at -20°C for long-term stability.
  • Available in small research quantities such as 10 MG for laboratory use.
Catalog No. 50-002-96698 Supplier Medchemexpress LLC Supplier No. HYB0803S110MG
Add to Cart
Add to Cart
Lumefantrine-d9 is a deuterium-labeled analog of lumefantrine used as an isotopically labeled internal standard and reference for analytical, pharmacokinetic, and metabolic studies. It enables accurate quantitation and method validation in mass spectrometry workflows.
  • Deuterium-labeled internal standard for lumefantrine assays.
  • Suitable for mass spectrometry based quantitation and tracer studies.
  • High purity and characterized molecular weight for reliable analytical results.
  • Stable as a powder; recommended storage at -20°C for long-term stability.
  • Available in small research quantities such as 10 MG for laboratory use.