Learn More
Apexbio Technology LLC LUMEFANTRINE-D9-5MG
SDP
Catalog No. 5000570277
Encompass_Preferred
Lumefantrine-d9 (CAS No 2477594-24-2) is a deuterated form of lumefantrine commonly used in the field of biomedical research particularly in antimalarial studies Originating from modifications to improve stability and metabolic profile this compound targets parasite growth by interfering with heme detoxification pathways vital to Plasmodium species It exhibits in vitro activity typically in the nanomolar to low micromolar range demonstrating efficacy in inhibiting parasite proliferation This makes it an ideal candidate for applications in various cell models and animal studies as well as in high-throughput drug screening programs aimed at discovering novel antimalarial agents Experimental concentrations typically depend on the specific research design ensuring flexibility in its application across diverse investigative setups
Catalog No. 50-005-70277 Supplier Apexbio Technology LLC Supplier No. SL10685
Lumefantrine-d9 (CAS No 2477594-24-2) is a deuterated form of lumefantrine commonly used in the field of biomedical research particularly in antimalarial studies Originating from modifications to improve stability and metabolic profile this compound targets parasite growth by interfering with heme detoxification pathways vital to Plasmodium species It exhibits in vitro activity typically in the nanomolar to low micromolar range demonstrating efficacy in inhibiting parasite proliferation This makes it an ideal candidate for applications in various cell models and animal studies as well as in high-throughput drug screening programs aimed at discovering novel antimalarial agents Experimental concentrations typically depend on the specific research design ensuring flexibility in its application across diverse investigative setups