Learn More
Medchemexpress LLC Lumefantrine-d9 (Benflumetol-d9) | 2477594-24-2 | 97.0% | 5 MG
SDP
Catalog No. 5000407515
Encompass_Preferred
Lumefantrine-d9 is a deuterium-labeled version of Lumefantrine, an antimalarial drug. It is commonly used in combination with Artemether, forming a combination therapy (AL) that serves as a first- and second-line antimalarial treatment.
  • Can be utilized as a tracer in research applications.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable isotope used for quantitation during drug development processes, with potential to influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07515 Supplier Medchemexpress LLC Supplier No. HYB0803S15MG
Lumefantrine-d9 is a deuterium-labeled version of Lumefantrine, an antimalarial drug. It is commonly used in combination with Artemether, forming a combination therapy (AL) that serves as a first- and second-line antimalarial treatment.
  • Can be utilized as a tracer in research applications.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable isotope used for quantitation during drug development processes, with potential to influence pharmacokinetic and metabolic profiles of drugs.