Learn More
Cayman Chemical Lumiflavine, Purity: ≥95%, CAS: 1088-56-8, 50 mg
SDP
Catalog No. 502981800
Encompass_Preferred
A structural analog of riboflavin; inhibits the uptake of radiolabeled riboflavin approximately 3-fold (at 25 µM) and 30-fold (at 10 µM) in Caco-2 cells and hRPE cells, respectively
Catalog No. 50-298-1800 Supplier Cayman Chemical Supplier No. 2064550 MG
A structural analog of riboflavin; inhibits the uptake of radiolabeled riboflavin approximately 3-fold (at 25 µM) and 30-fold (at 10 µM) in Caco-2 cells and hRPE cells, respectively