Learn More
Apexbio Technology LLC Luminescent ATP Cell Viability Assay Kit I(Synonyms: ATP Luminescent Cell Viability Assay, Luminescent ATP Assay Kit), 100T.
SDP
Catalog No. 5000061081
Encompass_Preferred
Luminescent ATP Cell Viability Assay Kit I uses thermostable firefly luciferase and purified luciferin substrate to quantitatively measure cellular ATP levels an indicator closely correlated with viable cell numbers and metabolic activity Upon reagent addition cells rapidly lyse to release intracellular ATP which interacts with luciferase and luciferin substrate generating a luminescent signal directly proportional to the number of viable cells The protocol does not require prior cell lysis steps offering a streamlined assay completed within 10 minutes This assay is suitable for detecting viability ranging from 10 to 30 000 cells and provides comparable results to common commercial ATP-based viability assays such as CellTiter-Glo
Catalog No. 50-000-61081 Supplier Apexbio Technology LLC Supplier No. K2041100
Luminescent ATP Cell Viability Assay Kit I uses thermostable firefly luciferase and purified luciferin substrate to quantitatively measure cellular ATP levels an indicator closely correlated with viable cell numbers and metabolic activity Upon reagent addition cells rapidly lyse to release intracellular ATP which interacts with luciferase and luciferin substrate generating a luminescent signal directly proportional to the number of viable cells The protocol does not require prior cell lysis steps offering a streamlined assay completed within 10 minutes This assay is suitable for detecting viability ranging from 10 to 30 000 cells and provides comparable results to common commercial ATP-based viability assays such as CellTiter-Glo