Learn More
Apexbio Technology LLC Luminescent ATP Cell Viability Assay Kit II(Synonyms: Luminescent ATP Viability Assay, ATP Luminescence Cell Viability Assay Kit), 1000T.
SDP
Catalog No. 5000061304
Encompass_Preferred
Luminescent ATP Cell Viability Assay Kit II assesses cell viability by quantitatively detecting intracellular adenosine triphosphate (ATP) an indicator proportional to metabolically active viable cells The assay utilizes thermostable firefly luciferase-mediated luminescence added reagents lyse cells releasing ATP which then reacts with supplied substrate under luciferase catalysis generating a luminescent signal directly proportional to live cell number This lyophilized formulation offers improved stability at -20 C storage short assay duration ( 10 min) enhanced sensitivity and linear quantification between approximately 10 to 30 000 cells facilitating biomedical assay applications
Catalog No. 50-000-61304 Supplier Apexbio Technology LLC Supplier No. K20421000
Luminescent ATP Cell Viability Assay Kit II assesses cell viability by quantitatively detecting intracellular adenosine triphosphate (ATP) an indicator proportional to metabolically active viable cells The assay utilizes thermostable firefly luciferase-mediated luminescence added reagents lyse cells releasing ATP which then reacts with supplied substrate under luciferase catalysis generating a luminescent signal directly proportional to live cell number This lyophilized formulation offers improved stability at -20 C storage short assay duration ( 10 min) enhanced sensitivity and linear quantification between approximately 10 to 30 000 cells facilitating biomedical assay applications