Learn More
Apexbio Technology LLC Luminescent ATP Detection Assay Kit(Synonyms: ATP Luminescence Assay Kit, ATP Bioluminescent Assay Kit, ATP Determination Kit), 200T.
SDP
Catalog No. 5000061174
Encompass_Preferred
Luminescent ATP Detection Assay Kit employs firefly luciferase-mediated bioluminescence for quantitative analysis of ATP concentration Luciferase catalyzes oxidation of the substrate D-luciferin in the presence of ATP resulting in emission of luminescence with intensity directly proportional to ATP levels This assay enables sensitive measurement within a linear detection range between 1 nM and 10 M ATP applicable for solution cell and tissue samples without complex extraction The detection procedure maintains stable luminescent signals for up to 30 min and samples remain compatible for further downstream analyses such as protein quantification SDS-PAGE and Western blotting
Catalog No. 50-000-61174 Supplier Apexbio Technology LLC Supplier No. K2040200
Luminescent ATP Detection Assay Kit employs firefly luciferase-mediated bioluminescence for quantitative analysis of ATP concentration Luciferase catalyzes oxidation of the substrate D-luciferin in the presence of ATP resulting in emission of luminescence with intensity directly proportional to ATP levels This assay enables sensitive measurement within a linear detection range between 1 nM and 10 M ATP applicable for solution cell and tissue samples without complex extraction The detection procedure maintains stable luminescent signals for up to 30 min and samples remain compatible for further downstream analyses such as protein quantification SDS-PAGE and Western blotting