Learn More
Hoefer, Inc LumiBlot Near-Field Chemi Imager
SDP
Supplier:  Hoefer, Inc LUMIBLOT
Encompass_Preferred
The LumiBlot Near-Field Chemi Imager combines ultra-high chemi sensitivity with next-generation near-field detection technology. Designed to overcome the limitations of traditional cooled CCD systems, LumiBlot delivers an improvement in sensitivity, quantitative performance, and workflow efficiency—empowering researchers to obtain faster, clearer, and more reliable blotting results.FeaturesUltra-SensitiveDetect even the weakest signals—100×more sensitive than traditional CCDs.Lightning-Fast ImagingGet results in just 200ms–1s. No more long waits.Wide Dynamic Range6 OD range captures strong and weak signals simultaneously with precision.OTA UpgradeUpgrade software online anytime to stay current.Multi-Device Connectionmonitors, laptops, tablets, and phones. ApplicationsWestern Blotting - Chemiluminescence imaging and quantitative analysisDot Blotting - Chemi imaging and quantitative analysis without electrophoresisSouthern and Northern Blotting
Catalog No. NC3942885