Learn More
ACROBiosystems Recombinant Protein;Human Lumican Protein;HEK293;1MG;LUM,LUM,Lumican,LDC,SLRR2D,KSPG lumican
Catalog No. 501658570
Encompass_Preferred
Recombinant Protein;Human Lumican Protein;1MG;Human Lumican, His Tag (LUM-H5227) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Asn 338 (Accession # NP_002336).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.5 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8570 Supplier ACROBiosystems Supplier No. LUMH52271MG
Recombinant Protein;Human Lumican Protein;1MG;Human Lumican, His Tag (LUM-H5227) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Asn 338 (Accession # NP_002336).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.5 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.