Learn More
Medchemexpress LLC Lumichrome | 50MG
SDP
Catalog No. 5000228720
Encompass_Preferred
Lumichrome a photodegradation product of Riboflavin is an endogenous compound in humans Lumichrome inhibits human lung cancer cell growth and induces apoptosis via a p53-dependent mechanism Lumichrom is the inhibitor for AKT/ -catenin signaling pathway[1][2]
Catalog No. 50-002-28720 Supplier Medchemexpress LLC Supplier No. HY11538550MG
Lumichrome a photodegradation product of Riboflavin is an endogenous compound in humans Lumichrome inhibits human lung cancer cell growth and induces apoptosis via a p53-dependent mechanism Lumichrom is the inhibitor for AKT/ -catenin signaling pathway[1][2]