Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Lumicyano Cyanoacrylate Latent Print Developer
SDP

Catalog No. 22110514 Shop All Mopec Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-110-514 1 g
22-110-445 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-110-514 Supplier Mopec Supplier No. FS505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for cyanoacrylate fuming to develop latent fingerprints that fluoresce directly after processing, reducing the need for an additional dye-staining step.

  • Available as a powder or solution
  • One-step process for showing fingerprints in fluorescence
  • Lumicyano technology is fully compatible with DNA analysis
  • Compatible with fuming chambers operating under standard fuming conditions
  • Hygrometry: 80%
  • Hotplate temperature: 250°F
  • Lumicyano is a forensic chemical reagent used to develop latent (invisible) fingerprints
  • Designed to reveal fingerprints with greater brightness and develop lightly detailed fingerprints under optimal conditions
  • Minimizes the risk of Over-gluing
  • Available in two configurations:
    • 1 g Powder / 20 g Solution
    • 5 g Powder / 100 g Solution
  • Allows double Identification fingerprint analysis followed by DNA analysis at one location

Specifications

Product Type Fingerprint Developer
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.