Learn More
Analytical Sales & Services LUMIDOX GEN2 24 WELL 18MM SPAC
Supplier:  Analytical Sales & Services LUM22418LA470
Encompass
LumiDox Gen2, 24 Well, 18mm Spacing 470-BLUE LED Array, with Lens Mat and Active Base
Catalog No. NC2076010